Please login to your online account to display your discounted pricing

anti-AU5 Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600461

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600461

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Formulation phosphate Buffered Saline (PBS)
Monoclonal or Polyclonal Polyclonal
Host Species Goat
Concentration 1.0mg/mL
Regulatory Status RUO
Format Affinity Purified
Immunogen Goats were immunized with AU5 (TDFYLK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Cross Reactivity Non-species specific
Quantity 0.1mg
Primary or Secondary Primary
Antigen AU5 Epitope Tag