Please login to your online account to display your discounted pricing

anti-ATRIP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100359SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100359SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 50% Glycerol
Gene Accession No. Q8WXE1
Gene ID (Entrez) 84126
Concentration 1mg/mL
Regulatory Status RUO
Test Specificity Human. Reactivity with intraprimates is unlikely owing to immunogen sequence divergence.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Immunogen A synthetic peptide derived from a sequence between amino acids 500-550 of human ATRIP [UniProt No. Q8WXE1]
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Antigen ATRIP
Format Affinity Purified
Host Species Rabbit
Cross Reactivity Human