Please login to your online account to display your discounted pricing

anti-ATRIP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100359

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100359

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Gene Accession No. Q8WXE1
Gene ID (Entrez) 84126
Concentration 1mg/mL
Formulation PBS, 50% Glycerol
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human. Reactivity with intraprimates is unlikely owing to immunogen sequence divergence.
Host Species Rabbit
Conjugate Unlabeled
Format Affinity Purified
Immunogen A synthetic peptide derived from a sequence between amino acids 500-550 of human ATRIP [UniProt No. Q8WXE1]
Quantity 0.1mL
Antigen ATRIP
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Cross Reactivity Human