Please login to your online account to display your discounted pricing

anti-ATRIP, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from a sequence between amino acids 500-550 of human ATRIP [UniProt No. Q8WXE1]
Antigen ATRIP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100359SS

 
novus biologicals canada
NB100359SS
Unlabeled 0.025mL Each for $109.00

NB100359

 
novus biologicals canada
NB100359
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from a sequence between amino acids 500-550 of human ATRIP [UniProt No. Q8WXE1]
Antigen ATRIP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Human. Reactivity with intraprimates is unlikely owing to immunogen sequence divergence.
Gene Accession No. Q8WXE1
Gene ID (Entrez) 84126
Formulation PBS, 50% Glycerol
Concentration 1mg/mL