Please login to your online account to display your discounted pricing

anti-ATPase Polyclonal, Thermo Scientific™

ATPase Polyclonal antibody PA1-18320 from Thermo Scientific™ specifically detects ATPase in Rat samples, and it is validated for Immunohistochemistry

$506.05 - $506.05

Specifications

Antigen ATPase
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-573

 
pierce biotechnology inc
PA118320
Unlabeled 100μL Each for $506.05

01-670-574

 
pierce biotechnology inc
PA118321
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen ATPase
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

ATPase Polyclonal antibody PA1-18320 from Thermo Scientific™ specifically detects ATPase in Rat samples, and it is validated for Immunohistochemistry


Host Species: Rabbit
Cross Reactivity: Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues C-D(195) D V H R S R H G L S L Q D Q(209) of human ATP13A2.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Probable cation-transporting ATPase 13A2, EC 3.6.3., ATP13A2

Gene Symbol: Atp13a2
Entrez GeneID: 362645
PA1-18320 detects ATPase in Rat samples. PA1-18320 has been successfully used in Immunohistochemistry procedures. PA1-18320 The immunogen is Synthetic peptide corresponding to residues C-D(195) D V H R S R H G L S L Q D Q(209) of human ATP13A2..