Please login to your online account to display your discounted pricing

anti-ATP5E, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142805SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142805SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Test Specificity Human, mouse, rat, porcine, bovine and Xenopus. Does not react with Drosophila.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Xenopus
Gene Accession No. P56381
Gene ID (Entrez) 514
Formulation PBS
Regulatory Status RUO
Antigen ATP5E
Quantity 0.025mL
Immunogen Synthetic peptide made to a sequence between amino acids 2-51 of human ATP5E. [Swiss-Prot No. P56381]
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting