Please login to your online account to display your discounted pricing

anti-ATP5E, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142805

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142805

  • / Each
 

Description & Specifications

Specifications

Antigen ATP5E
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Xenopus
Format Affinity Purified
Formulation PBS
Gene Accession No. P56381
Host Species Rabbit
Immunogen Synthetic peptide made to a sequence between amino acids 2-51 of human ATP5E. [Swiss-Prot No. P56381]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 514
Test Specificity Human, mouse, rat, porcine, bovine and Xenopus. Does not react with Drosophila.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rat
Target Species Xenopus