Please login to your online account to display your discounted pricing

anti-ATP5E, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a portion of Drosophila ATP5E (within residues 1-50). [Swiss-Prot No. Q9VXN2]
Antigen ATP5E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142813SS

 
novus biologicals canada
NBP142813SS
Unlabeled 0.025mL Each for $109.00

NBP142813

 
novus biologicals canada
NBP142813
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a portion of Drosophila ATP5E (within residues 1-50). [Swiss-Prot No. Q9VXN2]
Antigen ATP5E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Drosophila. Does not react with human.
Gene ID (Entrez) 514
Formulation PBS
Concentration 1mg/mL