Please login to your online account to display your discounted pricing

anti-ATP5E, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a sequence between amino acids 2-51 of human ATP5E. [Swiss-Prot No. P56381]
Antigen ATP5E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142805

 
novus biologicals canada
NBP142805
Unlabeled 0.1mL Each for $329.00

NBP142805SS

 
novus biologicals canada
NBP142805SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a sequence between amino acids 2-51 of human ATP5E. [Swiss-Prot No. P56381]
Antigen ATP5E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Human, mouse, rat, porcine, bovine and Xenopus. Does not react with Drosophila.
Gene Accession No. P56381
Gene ID (Entrez) 514
Formulation PBS
Concentration 1mg/mL