Please login to your online account to display your discounted pricing

anti-ATP5E, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142813

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142813

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Cross Reactivity Drosophila
Concentration 1mg/mL
Antigen ATP5E
Test Specificity Drosophila. Does not react with human.
Gene ID (Entrez) 514
Formulation PBS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen Synthetic peptide made to a portion of Drosophila ATP5E (within residues 1-50). [Swiss-Prot No. Q9VXN2]
Regulatory Status RUO
Conjugate Unlabeled
Quantity 0.1mL
Host Species Rabbit