Please login to your online account to display your discounted pricing

anti-ATG9A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11056893

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11056893

  • / Each
 

Description & Specifications

Specifications

Antigen ATG9A
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q7Z3C6
Host Species Rabbit
Immunogen A synthetic peptide made to a region within the C-terminus of human ATG9A (between residues 750-839). [Swiss-Prot No. Q7Z3C6].
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 79065
Target Species Bovine
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat