Please login to your online account to display your discounted pricing

anti-ATG7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224682SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224682SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Antigen ATG7
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O95352
Gene ID (Entrez) 10533
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and cow, 94% homologous in monkey and 83% homologous in chicken.
Immunogen A portion of amino acids 550-600 of human APG7L was used as the immunogen.
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Format Purified