Please login to your online account to display your discounted pricing

anti-ATG7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224682

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224682

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O95352
Gene ID (Entrez) 10533
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and cow, 94% homologous in monkey and 83% homologous in chicken.
Format Purified
Concentration 0.5mg/mL
Conjugate Unlabeled
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Cross Reactivity Human
Antigen ATG7
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Immunogen A portion of amino acids 550-600 of human APG7L was used as the immunogen.