Please login to your online account to display your discounted pricing

anti-ATG7, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 550-600 of human APG7L was used as the immunogen.
Antigen ATG7
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224682

 
novus biologicals canada
NBP224682
Unlabeled 0.1mg Each for $257.41

NBP224682SS

 
novus biologicals canada
NBP224682SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 550-600 of human APG7L was used as the immunogen.
Antigen ATG7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and cow, 94% homologous in monkey and 83% homologous in chicken.
Gene Accession No. O95352
Gene ID (Entrez) 10533
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL