missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ATG5, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human APG5L was used as the immunogen.
Antigen ATG5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224389SS Novus Biologicals Canada
NBP224389SS
0.025mg Each for $139.00
NBP224389 Novus Biologicals Canada
NBP224389
0.1mg Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
ATG5
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, opossum, horse, cow, dog, monkey, mouse, rat and chimpanzee and 95% homologous in chicken, 80% homologous in xenopus and 75% homologous in zebra fish.
100μg in 200μL PBS containing 0.2% gelatin
A portion of amino acid 1-50 of human APG5L was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
9474
0.5mg/mL