missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ATG5, Polyclonal, Novus Biologicals™

$164.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region of the human ATG5 protein (within residues 1-50) [Swiss-Prot Q9H1Y0]
Antigen ATG5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11053818T Novus Biologicals Canada
NB11053818SS
0.025mL Each for $164.00
NB11053818 Novus Biologicals Canada
NB11053818
0.1mL Each for $384.00
Specifications Description & Specifications

Specifications

Rabbit
ATG5
Affinity Purified
RUO
This is selective for the full-length and calpain cleaved isoform proteins. The short isoform is missing a.a. 1-79. The calpain cleaved form of ATG5 is missing a.a. 195-275.
9474
1.16mg/mL
A synthetic peptide made to an N-terminal region of the human ATG5 protein (within residues 1-50) [Swiss-Prot Q9H1Y0]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Simple Western,Western Blot
Q9H1Y0
PBS, 30% glycerol