Please login to your online account to display your discounted pricing

anti-ATG4B, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 350-400 of human APG4B was used as the immunogen.
Antigen ATG4B
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224709

 
novus biologicals canada
NBP224709
Unlabeled 0.1mg Each for $259.02

NBP224709SS

 
novus biologicals canada
NBP224709SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 350-400 of human APG4B was used as the immunogen.
Antigen ATG4B
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, monkey, rat, dog and orangutan, 94% homologous in opossum and mouse, 88% in zebrafish, pig, and catfish.
Gene ID (Entrez) 23192
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL