Please login to your online account to display your discounted pricing

anti-APG3 Polyclonal, Thermo Scientific™

APG3 Polyclonal Ab PA1-18297 from Thermo Scientific™ specifically detects APG3 in Hu and Rt samples, and it is validated for IF, IHC and WB

$506.05 - $506.05

Specifications

Antigen APG3
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-550

 
pierce biotechnology inc
PA118297
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen APG3
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal residues of human ATG3.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

APG3 Polyclonal Ab PA1-18297 from Thermo Scientific™ specifically detects APG3 in Hu and Rt samples, and it is validated for IF, IHC and WB

Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome or vacuole. Human ATG3 displays the same enzymatic characteristics in vitro as yeast Apg3, a protein-conjugating enzyme essential for autophagy (Tanida et al., 2002 [PubMed 11825910]).
Host Species: Rabbit
Cross Reactivity: Human, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the N-terminal residues of human ATG3.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Autophagy-related protein 3, APG3, APG3-like, hApg3, Protein PC3-96 , Autophagy-related E2-like conjugation enzyme ATG3, APG3, APG3L
Uniprot Accession: Q6AZ50, Q9NT62
Gene Symbol: ATG3
Entrez GeneID: 171415, 64422
PA1-18297 detects APG3 in Human and Rat samples. PA1-18297 has been successfully used in Immunofluorescence, Immunohistochemistry and Western Blot procedures. PA1-18297 The immunogen is Synthetic peptide corresponding to the N-terminal residues of human ATG3..