Please login to your online account to display your discounted pricing

anti-ATG16L1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224730SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224730SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 55054
Concentration 0.5mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey,95% homologous in dog, horse and mouse,91% homologous incow and rat.
Conjugate Unlabeled
Immunogen A portion of amino acids 25-75 of human APG16L was used as the immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Antigen ATG16L1
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO