Please login to your online account to display your discounted pricing

anti-ATG16L1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224730

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224730

  • / Each
 

Description & Specifications

Specifications

Antigen ATG16L1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 25-75 of human APG16L was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55054
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey,95% homologous in dog, horse and mouse,91% homologous incow and rat.
Target Species Human
Target Species Murine
Target Species Primate