Please login to your online account to display your discounted pricing

anti-ATG16L1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 25-75 of human APG16L was used as the immunogen.
Antigen ATG16L1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224730

 
novus biologicals canada
NBP224730
Unlabeled 0.1mg Each for $255.80

NBP224730SS

 
novus biologicals canada
NBP224730SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 25-75 of human APG16L was used as the immunogen.
Antigen ATG16L1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey,95% homologous in dog, horse and mouse,91% homologous incow and rat.
Gene ID (Entrez) 55054
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL