Please login to your online account to display your discounted pricing

anti-ATG16L1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of mouse ATG16L1. [Swiss-Prot No. Q8C0J2]
Antigen ATG16L1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11082384

 
novus biologicals canada
NB11082384
Unlabeled 0.1mL Each for $309.05

NB11082384T

 
novus biologicals canada
NB11082384SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of mouse ATG16L1. [Swiss-Prot No. Q8C0J2]
Antigen ATG16L1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 55054
Formulation PBS, 30% glycerol
Concentration 0.65mg/mL