Please login to your online account to display your discounted pricing

anti-ATF3, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt No. P18847]
Antigen ATF3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP102935SS

 
novus biologicals canada
NBP102935SS
Unlabeled 0.025mL Each for $144.63

NBP102935

 
novus biologicals canada
NBP102935
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt No. P18847]
Antigen ATF3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human, mouse and rat. Predicted to react with most mammalian species based on sequence homology.
Gene Accession No. P18847
Gene ID (Entrez) 467
Formulation PBS, 30% glycerol
Concentration 1.27mg/mL