Please login to your online account to display your discounted pricing

anti-ATF2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 13-27.
Antigen ATF2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056331

 
novus biologicals canada
NB10056331
Unlabeled 0.1mg Each for $255.80

NB10056331T

 
novus biologicals canada
NB10056331SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 13-27.
Antigen ATF2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Gene Accession No. NP_001871
Gene ID (Entrez) 1386
Formulation PBS
Concentration 0.5mg/mL