Please login to your online account to display your discounted pricing

anti-Aspartate Aminotransferase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP154778SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP154778SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P17174
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2805
Regulatory Status RUO
Cross Reactivity Equine
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Formulation PBS and 2% Sucrose Lyophilized with the antibody.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Quantity 0.02mg
Applications Western Blotting
Host Species Rabbit
Antigen Aspartate Aminotransferase
Conjugate Unlabeled
Immunogen Synthetic peptides corresponding to GOT1(glutamic-oxaloacetic transaminase 1
Format Purified