Please login to your online account to display your discounted pricing

anti-Aspartate Aminotransferase, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptides corresponding to GOT1(glutamic-oxaloacetic transaminase 1
Antigen Aspartate Aminotransferase
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP154778SS

 
novus biologicals canada
NBP154778SS
Unlabeled 0.02mg Each for $139.00

NBP154778

 
novus biologicals canada
NBP154778
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptides corresponding to GOT1(glutamic-oxaloacetic transaminase 1
Antigen Aspartate Aminotransferase
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. P17174
Gene ID (Entrez) 2805
Formulation PBS and 2% Sucrose Lyophilized with the antibody.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.