Please login to your online account to display your discounted pricing

anti-ASC/TMS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056365SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056365T

  • / Each
 

Description & Specifications

Specifications

Antigen ASC/TMS1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.2% gelatin
Gene Accession No. Q9ULZ3
Host Species Rabbit
Immunogen This polyclonal antibody was raised against a mixture of synthetic peptides corresponding to amino acid residues 31-46 and 94-110 of human TMS1.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 29108
Target Species Human