Please login to your online account to display your discounted pricing

anti-ASC/TMS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056075

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056075

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 29108
Conjugate Unlabeled
Quantity 0.05mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity ASC .
Immunogen A synthetic peptide corresponding to amino acids 83-102 (GQLQAATHQGSGAAPAGIQA) of human ASC/TMS1 was used as immunogen; GenBank no. NP_037390.2
Primary or Secondary Primary
Regulatory Status RUO
Format Antiserum
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Antigen ASC/TMS1
Host Species Rabbit