Please login to your online account to display your discounted pricing

anti-ASC/TMS1, Polyclonal, Novus Biologicals™

$139.00 - $139.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [UniProt Q9ULZ3]
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178978SS

 
novus biologicals canada
NBP178978SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [UniProt Q9ULZ3]
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q9ULZ3
Gene ID (Entrez) 29108
Formulation PBS, 30% glycerol
Concentration 1.12mg/mL