missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ASC/TMS1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a mixture of synthetic peptides corresponding to amino acid residues 31-46 and 94-110 of human TMS1.
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056365T Novus Biologicals Canada
NB10056365SS
0.025mg Each for $139.00
NB10056365 Novus Biologicals Canada
NB10056365
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
ASC/TMS1
Purified
RUO
Q9ULZ3
0.2% gelatin
This polyclonal antibody was raised against a mixture of synthetic peptides corresponding to amino acid residues 31-46 and 94-110 of human TMS1.
Polyclonal
Primary
Western Blot
29108
0.5mg/mL