Please login to your online account to display your discounted pricing

anti-ASC/TMS1, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human ASC/TMS1 protein (between residues 1-50) [UniProt Q9ULZ3]
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178977SS

 
novus biologicals canada
NBP178977SS
Unlabeled 0.025mL Each for $139.00

NBP178977

 
novus biologicals canada
NBP178977
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human ASC/TMS1 protein (between residues 1-50) [UniProt Q9ULZ3]
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Test Specificity Reactivity with Rat reported in PMID 24464748 .
Gene Accession No. Q9ULZ3
Gene ID (Entrez) 29108
Formulation PBS, 30% glycerol
Concentration 1.08mg/mL