Please login to your online account to display your discounted pricing

anti-ASB3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056645

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056645

  • / Each
 

Description & Specifications

Specifications

Antigen ASB3
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation PBS containing 0.1mg/mL BSA.
Gene Accession No. Q9Y575
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 63-75 and 166-180 of human Asb-3 (NP_665862).
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51130
Test Specificity Cross reacts with Human. It does not cross-react with Mouse or Rat Asb-3 .
Target Species Human