missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ASB2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against synthetic peptides containing amino acids 75-93 and 348-368 of human Asb-2 protein. Human spleen lysate can be used as a positive control. It should cross react with mouse and rat Asb-2
Antigen ASB2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056638 Novus Biologicals Canada
NB10056638
0.2mL Each for $255.80
NB10056638T Novus Biologicals Canada
NB10056638SS
0.05mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
ASB2
Antisera
RUO
Cross reacts with Human. Predicted to react with Mouse and Rat.
51676
0.5mg/mL
This polyclonal antibody was raised against synthetic peptides containing amino acids 75-93 and 348-368 of human Asb-2 protein. Human spleen lysate can be used as a positive control. It should cross react with mouse and rat Asb-2
Polyclonal
Primary
Western Blot
Q96Q27
PBS containing 0.05% BSA