Please login to your online account to display your discounted pricing

anti-ASB11, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056640

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056640

  • / Each
 

Description & Specifications

Specifications

Antigen ASB11
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.1mg/mL BSA.
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 40-51 and 250-265 of human Asb-11.
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 140456
Target Species Human