missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ASAP1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 962-999 of mouse ASAP1 were used as the immunogen.
Antigen ASAP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224643SS Novus Biologicals Canada
NBP224643SS
0.025mg Each for $109.00
NBP224643 Novus Biologicals Canada
NBP224643
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
ASAP1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in rat, 92% homologous in human and mouse and 85% in chicken.
100μg in 200μL PBS containing 0.05% BSA
Within the range of amino acids 962-999 of mouse ASAP1 were used as the immunogen.
Polyclonal
Primary
Western Blot
50807
0.5mg/mL