Please login to your online account to display your discounted pricing

anti-CD3, Clone: 145-2C11, Novus Biologicals™

$139.00 - $332.66

Specifications

Clone 145-2C11
Host Species Armenian Hamster
Isotype IgG
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Antigen CD3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP230151

 
novus biologicals canada
NBP230151
Unlabeled 0.1mg Each for $332.66

NBP230151SS

 
novus biologicals canada
NBP230151SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 145-2C11
Host Species Armenian Hamster
Isotype IgG
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Antigen CD3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene ID (Entrez) 916
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL