Please login to your online account to display your discounted pricing

anti-CD3, Clone: 145-2C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230151SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230151SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Host Species Armenian Hamster
Antigen CD3
Format Purified
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Applications Flow Cytometry
Isotype IgG
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Gene ID (Entrez) 916
Concentration 0.5mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Clone 145-2C11
Regulatory Status RUO