Please login to your online account to display your discounted pricing

anti-CD3, Clone: 145-2C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230151

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230151

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Mouse
Format Purified
Applications Flow Cytometry
Quantity 0.1mg
Clone 145-2C11
Gene ID (Entrez) 916
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen CD3
Host Species Armenian Hamster
Isotype IgG
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37