Please login to your online account to display your discounted pricing

anti-CD11c, Clone: AP-MAB0806, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11097871SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11097871T

  • / Each
 

Description & Specifications

Specifications

Antigen CD11c
Format Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Armenian Hamster immunized with mouse lymphocytes.
Quantity 0.04mg
Host Species Armenian Hamster
Isotype IgG
Clone AP-MAB0806
Regulatory Status RUO
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in phosphate-buffered saline (PBS).
Gene ID (Entrez) 3687
Monoclonal or Polyclonal Monoclonal
Reconstitution Reconstitute the antibody with 400μL sterile PBS (final concentration 0.5mg/mL).
Cross Reactivity Mouse