Please login to your online account to display your discounted pricing

anti-CD11c, Clone: AP-MAB0806, Novus Biologicals™

$661.49 - $661.49

Specifications

Clone AP-MAB0806
Host Species Armenian Hamster
Isotype IgG
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Armenian Hamster immunized with mouse lymphocytes.
Antigen CD11c
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11097871

 
novus biologicals canada
NB11097871
Unlabeled 0.2mg Each for $661.49
Description & Specifications

Specifications

Clone AP-MAB0806
Host Species Armenian Hamster
Isotype IgG
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Armenian Hamster immunized with mouse lymphocytes.
Antigen CD11c
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Gene ID (Entrez) 3687
Formulation Lyophilized from a 0.2μm filtered solution in phosphate-buffered saline (PBS).
Reconstitution Reconstitute the antibody with 400μL sterile PBS (final concentration 0.5mg/mL).