Please login to your online account to display your discounted pricing

anti-ARID5B, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP224657SS

 View all options for this product


For Research Use Only

Catalog No. NBP224657SS


Description & Specifications


Antigen ARID5B
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q14865
Host Species Rabbit
Immunogen A portion of amino acids 500-550 of MRF2 (ARID5B) was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 84159
Test Specificity The amino acid sequence used as immunogen for the SOX2 antibody is 100% homologous in human, pig, platypus, salmon, chimpanzee, cat, mouse, rhesus monkey, Xenopus, zebrafish, chicken and Tetradon and 94% homologous in rat, Mouse and opossum.