Please login to your online account to display your discounted pricing

anti-ARID5B, Polyclonal, Novus Biologicals™

$106.00 - $321.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of MRF2 (ARID5B) was used as the immunogen for the antibody.
Antigen ARID5B
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224657SS

 
novus biologicals canada
NBP224657SS
Unlabeled 0.025mg Each for $106.00

NBP224657

 
novus biologicals canada
NBP224657
Unlabeled 0.1mg Each for $321.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of MRF2 (ARID5B) was used as the immunogen for the antibody.
Antigen ARID5B
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for the SOX2 antibody is 100% homologous in human, pig, platypus, salmon, chimpanzee, cat, mouse, rhesus monkey, Xenopus, zebrafish, chicken and Tetradon and 94% homologous in rat, Mouse and opossum.
Gene Accession No. Q14865
Gene ID (Entrez) 84159
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL