Please login to your online account to display your discounted pricing

anti-ARHGAP1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of human RhoGAP1 was used as the immunogen
Antigen ARHGAP1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224707

 
novus biologicals canada
NBP224707
Unlabeled 0.1mg Each for $255.80

NBP224707SS

 
novus biologicals canada
NBP224707SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of human RhoGAP1 was used as the immunogen
Antigen ARHGAP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat (each having multiple isoforms), 94% homologous in bovine, 88% homologous in chicken, 76% homologous in Xenopus, and 70% homologous in zebrafish.
Gene Accession No. Q07960
Gene ID (Entrez) 392
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL