Please login to your online account to display your discounted pricing

anti-Arg3.1 Polyclonal, Thermo Scientific™

Arg3.1 Polyclonal antibody PA1-18301 from Thermo Scientific™ specifically detects Arg3.1 in Human and Rat samples, and it is validated for Immunohistochemistry

$543.75 - $543.75

Specifications

Antigen Arg3.1
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-554

 
pierce biotechnology inc
PA118301
Unlabeled 100μL Each for $543.75

01-670-555

 
pierce biotechnology inc
PA118302
Unlabeled 500μg Each for $543.75
Description & Specifications

Specifications

Antigen Arg3.1
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues of human Arg3.1.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Arg3.1 Polyclonal antibody PA1-18301 from Thermo Scientific™ specifically detects Arg3.1 in Human and Rat samples, and it is validated for Immunohistochemistry

Arc (also termed activity-regulated cytoskeleton-associated protein or Arg3.1), is an effector immediate early gene whose upregulation has been demonstrated during events of synaptic plasticity. Arg3.1 expression is detectable in neuronal cell bodies and dendrites in the brain regions including striatum and cortex hippocampus, hypothalamus, amygdala.
Host Species: Rabbit
Cross Reactivity: Human, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the C-terminal residues of human Arg3.1.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: ARC, Growth factor ARC
Uniprot Accession: Q63053, Q7LC44
Gene Symbol: ARC
Entrez GeneID: 23237, 54323
PA1-18301 detects Arg3.1 in Human and Rat samples. PA1-18301 has been successfully used in Immunohistochemistry procedures. PA1-18301 The immunogen is Synthetic peptide corresponding to the C-terminal residues of human Arg3.1..