Please login to your online account to display your discounted pricing

anti-ARFGAP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224605SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224605SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 55738
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, horse, dog and monkey and 97% homologous in rat and opossum.
Antigen ARFGAP1
Gene Accession No. Q8N6T3
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Western Blotting
Host Species Rabbit
Immunogen Within the range of amino acids 123-183 of human ArfGAP1 were used as the immunogen.
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Regulatory Status RUO
Format Purified