Please login to your online account to display your discounted pricing

anti-ARFGAP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224605

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224605

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, horse, dog and monkey and 97% homologous in rat and opossum.
Format Purified
Gene Accession No. Q8N6T3
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 55738
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Within the range of amino acids 123-183 of human ArfGAP1 were used as the immunogen.
Primary or Secondary Primary
Applications Western Blotting
Host Species Rabbit
Quantity 0.1mg
Antigen ARFGAP1
Regulatory Status RUO