Please login to your online account to display your discounted pricing

anti-ARFGAP1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 123-183 of human ArfGAP1 were used as the immunogen.
Antigen ARFGAP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224605

 
novus biologicals canada
NBP224605
Unlabeled 0.1mg Each for $257.41

NBP224605SS

 
novus biologicals canada
NBP224605SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 123-183 of human ArfGAP1 were used as the immunogen.
Antigen ARFGAP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, horse, dog and monkey and 97% homologous in rat and opossum.
Gene Accession No. Q8N6T3
Gene ID (Entrez) 55738
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL