Please login to your online account to display your discounted pricing

anti-Apolipoprotein E R2/ApoE R2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt No. Q14114]
Antigen Apolipoprotein E R2/ApoE R2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002216

 
novus biologicals canada
NB1002216
Unlabeled 0.1mL Each for $329.00

NB1002216SS

 
novus biologicals canada
NB1002216SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt No. Q14114]
Antigen Apolipoprotein E R2/ApoE R2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, chicken and mouse. Immunogen has 95% homology to bovine.
Gene Accession No. Q14114
Gene ID (Entrez) 7804
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL