missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Apolipoprotein B Goat anti-Human, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AB742
Description
Anti-Apolipoprotein B Antibody is an antibody against Apolipoprotein B for use in immunoprecipitation and western blotting.Specifications
Apolipoprotein B | |
Polyclonal | |
Defibrinated, delipidized and absorbed by solid phase chromatography as required. Goat antisera in 0.05M Tris-HCl pH 7.5, 0.5M NaCl with 0.1% sodium azide. | |
Goat | |
1 mL | |
NM_000384.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Immunoprecipitation, Western Blot | |
Unconjugated | |
APOB; apoB-48; FLDB; apoB-100 | |
Purified human Apolipoprotein B. | |
Primary | |
Human | |
Serum |
For Research Use Only