Please login to your online account to display your discounted pricing

anti-APH1A, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-145 of human APH1A was used as the immunogen.
Antigen APH1A
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227176

 
novus biologicals canada
NBP227176
Unlabeled 0.1mg Each for $257.41

NBP227176SS

 
novus biologicals canada
NBP227176SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-145 of human APH1A was used as the immunogen.
Antigen APH1A
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, mouse, cow and dog, 94% homologous in rat and 88% homologous in chicken.
Gene ID (Entrez) 51107
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL